MilliporeSigma
Competition between Bending and Internal Pressure Governs the Mechanics of Fluid Nanovesicles.
ACS nano (2017-03-09)
Daan Vorselen, Fred C MacKintosh, Wouter H Roos, Gijs J L Wuite
RÉSUMÉ
Nanovesicles (∼100 nm) are ubiquitous in cell biology and an important vector for drug delivery. Mechanical properties of vesicles are known to influence cellular uptake, but the mechanism by which deformation dynamics affect internalization is poorly understood. This is partly due to the fact that experimental studies of the mechanics of such vesicles remain challenging, particularly at the nanometer scale where appropriate theoretical models have also been lacking. Here, we probe the mechanical properties of nanoscale liposomes using atomic force microscopy (AFM) indentation. The mechanical response of the nanovesicles shows initial linear behavior and subsequent flattening corresponding to inward tether formation. We derive a quantitative model, including the competing effects of internal pressure and membrane bending, that corresponds well to these experimental observations. Our results are consistent with a bending modulus of the lipid bilayer of ∼14k
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
L-α-phosphatidylcholine, egg yolk, Type XI-E, 100 mg/mL in chloroform, ≥99%, solution