MilliporeSigma
Association of pyruvate dehydrogenase kinase 4 gene polymorphisms with type 2 diabetes and metabolic syndrome.
Diabetes research and clinical practice (2011-10-25)
Seong-Su Moon, Jung-Eun Lee, Young-Sil Lee, Su-Won Kim, Nam Ho Jeoung, In-Kyu Lee, Jung-Guk Kim
RÉSUMÉ
Pyruvate dehydrogenase kinase 4 (PDK4) plays a crucial role in glucose utilization and lipid metabolism by regulating the pyruvate dehydrogenase complex (PDC) and is an emerging therapeutic target for type 2 diabetes. To date, no study has specifically examined the relationship between PDK4 gene polymorphisms and type 2 diabetes or metabolic syndrome. The association of common single nucleotide polymorphisms (SNPs) was examined in PDK4 [-208A/G (rs10085637), IVS3+192C/T (rs3779478), IVS6+31A/G (rs2301630), IVS7+514A/G (rs12668651), IVS10+75C/T (rs10247649)] with type 2 diabetes and metabolic syndrome in 651 Korean subjects with type 2 diabetes and 350 nondiabetic Korean subjects. The association of these SNPs with clinical parameters related to metabolic syndromes including obesity, hyperglycemia, hypertension, and dyslipidemia was also examined. No significant association was found between the studied SNPs and type 2 diabetes, metabolic syndrome, or clinical parameters. The PDK4 gene haplotype ACAGC showed a modest association with type 2 diabetes. However, the significance of this association was lost after considering for multiple comparisons. PDK4 polymorphisms may not be associated with type 2 diabetes or metabolic syndrome. Further studies utilizing a larger study population are required to confirm these results.