MilliporeSigma
  • Silencing of advanced glycosylation and glycosylation and product-specific receptor (RAGE) inhibits the metastasis and growth of non-small cell lung cancer.
Silencing of advanced glycosylation and glycosylation and product-specific receptor (RAGE) inhibits the metastasis and growth of non-small cell lung cancer.
American journal of translational research (2017-07-04)
Yan Xia Yu, Wen Chong Pan, Yu Feng Cheng
RÉSUMÉ
Non-small cell lung cancer (NSCLC) constitutes the main cases of lung cancer and is the world's most common and lethal cancer owing to regional invasion or distant metastasis. Growing morbidity and lethality demonstrates that valid molecular target in management of NSCLC metastasis is still absence. The receptor of advanced glycation end-products (RAGE) has been identified as an oncogenic gene and appears to promote the growth and metastasis of various cancers. Here, we investigated if RAGE targeted by RNA interference (RNAi) might have certain effect on the restraint of the growth of NSCLC and tumor metastasis. Wound healing and Transwell invasion assays indicated that RAGE favored the metastatic capabilities of NSCLC H1975 cells. Besides, soft-agar colony assay revealed that silencing RAGE significantly blocked colony-forming capability of H1975 cells in vitro. Furthermore, we observed that RAGE participated in H1975 cells growth, metastasis and epithelial-mesenchymal transition (EMT) by regulating interdict crux intracellular signaling pathways, including phosphatidylinositol-3 kinase/serine-threonine kinase (PI3K/AKT) and V-Ki-ras2 kirsten rat sarcoma viral oncogene homolog/RAF proto-oncogene serine/threonine-protein kinase (KRAS/RAF-1). In xenograft model, significantly reduction intumor growth and Ki67 expression was demonstrated in nude mice inoculation with RAGE down-regulation H1975 cells. To conclude, our study demonstrated that RAGE played a crucial role in the metastasis and growth of NSCLC by regulating PI3K/AKT and KRAS/RAF-1 signaling pathways, thereby might be a promising therapeutic target for NSCLC.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
MISSION® esiRNA, targeting human MOK