MilliporeSigma
Bitterness in sugar: O-GlcNAcylation aggravates pre-B acute lymphocytic leukemia through glycolysis via the PI3K/Akt/c-Myc pathway.
American journal of cancer research (2017-07-04)
Bing Zhang, Panpan Zhou, Xue Li, Qing Shi, Dong Li, Xiuli Ju
RÉSUMÉ
Abnormal cellular energetics has emerged as a hallmark of cancer cells. Deregulating aerobic glycolysis can alter multiple metabolic and signaling pathways in cancer cells, and trigger unlimited growth and proliferation. Accumulating evidence suggests that elevated levels of protein modification with β-N-acetylglucosamine (O-GlcNAcylation) along with dysregulation of O-GlcNAc transferase (OGT) and/or O-GlcNAcase (OGA) levels may act as a nutrient sensor in cancer cells. However, the underlying mechanism of O-GlcNAcylation and the relationship between O-GlcNAcylation and glycolysis are largely unknown in pre-B acute lymphocytic leukemia (pre-B-ALL). In this study, CD19
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
MISSION® esiRNA, targeting human OGT