MilliporeSigma
Methylation regulates HEY1 expression in glioblastoma.
Oncotarget (2017-06-03)
Andrew J Tsung, Maheedhara R Guda, Swapna Asuthkar, Collin M Labak, Ian J Purvis, Yining Lu, Neha Jain, Sarah E Bach, Durbaka V R Prasad, Kiran K Velpula
RÉSUMÉ
Glioblastoma (GBM) remains one of the most lethal and difficult-to-treat cancers of the central nervous system. The poor prognosis in GBM patients is due in part to its resistance to available treatments, which calls for identifying novel molecular therapeutic targets. In this study, we identified a mediator of Notch signaling, HEY1, whose methylation status contributes to the pathogenesis of GBM. Datamining studies, immunohistochemistry and immunoblot analysis showed that HEY1 is highly expressed in GBM patient specimens. Since methylation status of HEY1 may control its expression, we conducted bisulphite sequencing on patient samples and found that the HEY1 promoter region was hypermethylated in normal brain when compared to GBM specimens. Treatment on 4910 and 5310 xenograft cell lines with sodium butyrate (NaB) significantly decreased HEY1 expression with a concomitant increase in DNMT1 expression, confirming that promoter methylation may regulate HEY1 expression in GBM. NaB treatment also induced apoptosis of GBM cells as measured by flow cytometric analysis. Further, silencing of HEY1 reduced invasion, migration and proliferation in 4910 and 5310 cells. Furthermore, immunoblot and q-PCR analysis showed the existence of a potential positive regulatory loop between HEY1 and p53. Additionally, transcription factor interaction array with HEY1 recombinant protein predicted a correlation with p53 and provided various bonafide targets of HEY1. Collectively, these studies suggest HEY1 may be an important predictive marker for GBM and potential target for future GBM therapy.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Caspase 9 Colorimetric Activity Assay Kit, LEHD, Caspase-9 Colorimetric Activity Assay Kits provide a simple & convenient means for assaying the activity of caspases that recognize the LEHD.
Sigma-Aldrich
Caspase 3 Colorimetric Activity Assay Kit, DEVD, Caspase-3 Colorimetric Activity Assay Kits provide a simple & convenient means for assaying the activity of caspases that recognize the sequence DEVD.
Sigma-Aldrich
MISSION® esiRNA, targeting human HEY1