MilliporeSigma
GWAS of clinically defined gout and subtypes identifies multiple susceptibility loci that include urate transporter genes.
Annals of the rheumatic diseases (2016-12-03)
Akiyoshi Nakayama, Hirofumi Nakaoka, Ken Yamamoto, Masayuki Sakiyama, Amara Shaukat, Yu Toyoda, Yukinori Okada, Yoichiro Kamatani, Takahiro Nakamura, Tappei Takada, Katsuhisa Inoue, Tomoya Yasujima, Hiroaki Yuasa, Yuko Shirahama, Hiroshi Nakashima, Seiko Shimizu, Toshihide Higashino, Yusuke Kawamura, Hiraku Ogata, Makoto Kawaguchi, Yasuyuki Ohkawa, Inaho Danjoh, Atsumi Tokumasu, Keiko Ooyama, Toshimitsu Ito, Takaaki Kondo, Kenji Wakai, Blanka Stiburkova, Karel Pavelka, Lisa K Stamp, Nicola Dalbeth, Yutaka Sakurai, Hiroshi Suzuki, Makoto Hosoyamada, Shin Fujimori, Takashi Yokoo, Tatsuo Hosoya, Ituro Inoue, Atsushi Takahashi, Michiaki Kubo, Hiroshi Ooyama, Toru Shimizu, Kimiyoshi Ichida, Nariyoshi Shinomiya, Tony R Merriman, Hirotaka Matsuo
RÉSUMÉ
A genome-wide association study (GWAS) of gout and its subtypes was performed to identify novel gout loci, including those that are subtype-specific. Putative causal association signals from a GWAS of 945 clinically defined gout cases and 1213 controls from Japanese males were replicated with 1396 cases and 1268 controls using a custom chip of 1961 single nucleotide polymorphisms (SNPs). We also first conducted GWASs of gout subtypes. Replication with Caucasian and New Zealand Polynesian samples was done to further validate the loci identified in this study. In addition to the five loci we reported previously, further susceptibility loci were identified at a genome-wide significance level (p<5.0×10 Our findings including novel gout risk loci provide further understanding of the molecular pathogenesis of gout and lead to a novel concept for the therapeutic target of gout/hyperuricaemia.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Anti-FAM35A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution