MilliporeSigma
Improving 10-deacetylbaccatin III-10-β-O-acetyltransferase catalytic fitness for Taxol production.
Nature communications (2017-05-19)
Bing-Juan Li, Hao Wang, Ting Gong, Jing-Jing Chen, Tian-Jiao Chen, Jin-Ling Yang, Ping Zhu
RÉSUMÉ
The natural concentration of the anticancer drug Taxol is about 0.02% in yew trees, whereas that of its analogue 7-β-xylosyl-10-deacetyltaxol is up to 0.5%. While this compound is not an intermediate in Taxol biosynthetic route, it can be converted into Taxol by de-glycosylation and acetylation. Here, we improve the catalytic efficiency of 10-deacetylbaccatin III-10-O-acetyltransferase (DBAT) of Taxus towards 10-deacetyltaxol, a de-glycosylated derivative of 7-β-xylosyl-10-deacetyltaxol to generate Taxol using mutagenesis. We generate a three-dimensional structure of DBAT and identify its active site using alanine scanning and design a double DBAT mutant (DBAT
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Baccatin III, ≥95% (HPLC)