MilliporeSigma
  • Self-Host Blue-Emitting Iridium Dendrimer Containing Bipolar Dendrons for Nondoped Electrophosphorescent Devices with Superior High-Brightness Performance.
Self-Host Blue-Emitting Iridium Dendrimer Containing Bipolar Dendrons for Nondoped Electrophosphorescent Devices with Superior High-Brightness Performance.
ACS applied materials & interfaces (2016-11-03)
Yang Wang, Yaoming Lu, Baoxiang Gao, Shumeng Wang, Junqiao Ding, Lixiang Wang, Xiabin Jing, Fosong Wang
RÉSUMÉ
A novel self-host blue-emitting iridium dendrimer, namely, B-CzPO, has been designed and synthesized via a postdendronization route, where a bipolar carbazole/triphenylphosphine oxide hybrid is selected as the peripheral dendron instead of the p-type oligocarbazole used in unipolar analogue B-CzG2. This structural modification can render B-CzPO with more balanced charge transportation relative to that of B-CzG2. As a result of the significantly reduced efficiency roll-off, the nondoped phosphorescent organic light-emitting diodes (PhOLEDs) of B-CzPO show a superior high-brightness performance, revealing a luminous efficiency of 21.2, 16.1, and 10.5 cd/A at 1000, 5000, and 10 000 cd/m
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
SPPO13