MilliporeSigma
  • Electrochemical immunosensor for simultaneous determination of interleukin-1 beta and tumor necrosis factor alpha in serum and saliva using dual screen printed electrodes modified with functionalized double-walled carbon nanotubes.
Electrochemical immunosensor for simultaneous determination of interleukin-1 beta and tumor necrosis factor alpha in serum and saliva using dual screen printed electrodes modified with functionalized double-walled carbon nanotubes.
Analytica chimica acta (2017-02-06)
E Sánchez-Tirado, C Salvo, A González-Cortés, P Yáñez-Sedeño, F Langa, J M Pingarrón
RÉSUMÉ
Dual screen-printed carbon electrodes modified with 4-carboxyphenyl-functionalized double-walled carbon nanotubes (HOOC-Phe-DWCNTs/SPCEs) have been used as scaffolds for the preparation of electrochemical immunosensors for the simultaneous determination of the cytokines Interleukin-1β (IL-1β) and factor necrosis tumor α (TNF-α). IL-1β. Capture antibodies were immobilized onto HOOC-Phe-DWCNTs/SPCEs in an oriented form making using the commercial polymeric coating Mix&Go™. Sandwich type immunoassays with amperometric signal amplification through the use of poly-HRP-streptavidin conjugates and H
MATÉRIAUX
Référence du produit
Marque
Description du produit
Supelco
Streptavidin−Peroxidase Polymer, Ultrasensitive
Sigma-Aldrich
Sera human, lyophilized powder (from clotted whole blood)