MilliporeSigma
  • A biophysical investigation on the binding of proflavine with human hemoglobin: Insights from spectroscopy, thermodynamics and AFM studies.
A biophysical investigation on the binding of proflavine with human hemoglobin: Insights from spectroscopy, thermodynamics and AFM studies.
Journal of photochemistry and photobiology. B, Biology (2016-10-22)
Anirban Basu, Gopinatha Suresh Kumar
RÉSUMÉ
Interaction of proflavine with hemoglobin (Hgb) was studied employing spectroscopy, calorimetry, and atomic force microscopy. The equilibrium constant was found to be of the order 10
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
3,6-Diaminoacridine hydrochloride, Dye content 95 %