MilliporeSigma
The tau tubulin kinases TTBK1/2 promote accumulation of pathological TDP-43.
PLoS genetics (2014-12-05)
Nicole F Liachko, Pamela J McMillan, Timothy J Strovas, Elaine Loomis, Lynne Greenup, Jill R Murrell, Bernardino Ghetti, Murray A Raskind, Thomas J Montine, Thomas D Bird, James B Leverenz, Brian C Kraemer
RÉSUMÉ
Pathological aggregates of phosphorylated TDP-43 characterize amyotrophic lateral sclerosis (ALS) and frontotemporal lobar degeneration (FTLD-TDP), two devastating groups of neurodegenerative disease. Kinase hyperactivity may be a consistent feature of ALS and FTLD-TDP, as phosphorylated TDP-43 is not observed in the absence of neurodegeneration. By examining changes in TDP-43 phosphorylation state, we have identified kinases controlling TDP-43 phosphorylation in a C. elegans model of ALS. In this kinome-wide survey, we identified homologs of the tau tubulin kinases 1 and 2 (TTBK1 and TTBK2), which were also identified in a prior screen for kinase modifiers of TDP-43 behavioral phenotypes. Using refined methodology, we demonstrate TTBK1 and TTBK2 directly phosphorylate TDP-43 in vitro and promote TDP-43 phosphorylation in mammalian cultured cells. TTBK1/2 overexpression drives phosphorylation and relocalization of TDP-43 from the nucleus to cytoplasmic inclusions reminiscent of neuropathologic changes in disease states. Furthermore, protein levels of TTBK1 and TTBK2 are increased in frontal cortex of FTLD-TDP patients, and TTBK1 and TTBK2 co-localize with TDP-43 inclusions in ALS spinal cord. These kinases may represent attractive targets for therapeutic intervention for TDP-43 proteinopathies such as ALS and FTLD-TDP.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Anti-TTBK1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution