MilliporeSigma
  • The BRAF Inhibitor Vemurafenib Activates Mitochondrial Metabolism and Inhibits Hyperpolarized Pyruvate-Lactate Exchange in BRAF-Mutant Human Melanoma Cells.
The BRAF Inhibitor Vemurafenib Activates Mitochondrial Metabolism and Inhibits Hyperpolarized Pyruvate-Lactate Exchange in BRAF-Mutant Human Melanoma Cells.
Molecular cancer therapeutics (2016-10-22)
Teresa Delgado-Goni, Maria Falck Miniotis, Slawomir Wantuch, Harold G Parkes, Richard Marais, Paul Workman, Martin O Leach, Mounia Beloueche-Babari
RÉSUMÉ
Understanding the impact of BRAF signaling inhibition in human melanoma on key disease mechanisms is important for developing biomarkers of therapeutic response and combination strategies to improve long-term disease control. This work investigates the downstream metabolic consequences of BRAF inhibition with vemurafenib, the molecular and biochemical processes that underpin them, their significance for antineoplastic activity, and potential as noninvasive imaging response biomarkers.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
VPC-13566, ≥98% (HPLC)