MilliporeSigma
  • Organocatalytic asymmetric robinson annulation of alpha,beta-unsaturated aldehydes: applications to the total synthesis of (+)-palitantin.
Organocatalytic asymmetric robinson annulation of alpha,beta-unsaturated aldehydes: applications to the total synthesis of (+)-palitantin.
The Journal of organic chemistry (2007-10-09)
Bor-Cherng Hong, Ming-Fun Wu, Hsing-Chang Tseng, Guo-Fong Huang, Cheng-Feng Su, Ju-Hsiou Liao
RÉSUMÉ
The highly enantioselective organocatalytic Robinson annulation of alpha,beta-unsaturated aldehydes was achieved, catalyzed by l-proline and trialkylamines and providing the formal [4 + 2] cycloaddition adducts. Additionally, in some examples in the catalysis with diarylpyrrolinol silyl ethers, the reactions afforded the [4 + 2] adducts with high enantioselectivity (>99.5% ee). The structure of the adduct, obtained from the reaction of 3-methylbut-2-enal and (E)-3-(2-nitrophenyl)acrylaldehyde, was confirmed by X-ray analysis. The absolute configurations of some [4 + 2] cycloadducts were investigated, and the methodology was applied in the synthesis of (+)-palitantin.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
(S)-(–)-α,α-Diphenyl-2-pyrrolidine methanol trimethylsilyl ether, 95%