MilliporeSigma
  • Lansoprazole, a novel benzimidazole proton pump inhibitor, and its related compounds have selective activity against Helicobacter pylori.
Lansoprazole, a novel benzimidazole proton pump inhibitor, and its related compounds have selective activity against Helicobacter pylori.
Antimicrobial agents and chemotherapy (1991-03-01)
T Iwahi, H Satoh, M Nakao, T Iwasaki, T Yamazaki, K Kubo, T Tamura, A Imada
RÉSUMÉ
The activities of various types of antiulcer agents against Helicobacter pylori (formerly called Campylobacter pylori) strains were determined by an agar dilution method. Among the compounds tested, two benzimidazole proton pump inhibitors, lansoprazole (AG-1749) and omeprazole, were found to have significant activities against this organism. The activity of lansoprazole was comparable to that of bismuth citrate, with MICs ranging from 3.13 to 12.5 micrograms/ml, and fourfold more potent than that of omeprazole. A major metabolite and two acid-converted rearrangement products of lansoprazole also exhibited good activities comparable or superior to that of the parent compound. Exposure to lansoprazole of H. pylori growing in a liquid medium led to an extensive loss of viability without a reduction in culture turbidity and produced an aberrant bacterial morphology characterized by the irregular constriction of cells and the collapse of cell surface structures. The antibacterial activity of lansoprazole and its related compounds was selective against H. pylori; common aerobic and anaerobic bacteria and Campylobacter jejuni were not inhibited by 100 micrograms/ml.