MilliporeSigma
Labelling saccharides with phenylhydrazine for electrospray and matrix-assisted laser desorption-ionization mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2003-07-26)
Erika Lattova, Hélène Perreault
RÉSUMÉ
A well-known reaction of carbonyl compounds with phenylhydrazine has been applied to saccharides, providing increased sensitivity for mass spectrometric (MS) and ultraviolet (UV) detection during high-performance liquid chromatographic (HPLC) separations. After a simple derivatization procedure for 1 h at 70 degrees C and purification of the reaction mixture from excess reagent by extraction, the sugar derivatives were characterized by direct injection or on-line HPLC/electrospray ionization (ESI) and by matrix-assisted laser desorption/ionization (MALDI) MS. Because no salts are used or produced upon reaction, this procedure is very simple and suitable for the tagging of saccharides. The reaction allows for on-target derivatization and products are very stable. The derivatization procedure has been applied to commercially-obtained small saccharides and standard N-linked oligosaccharides. Lastly, hen ovalbumin N-glycans were detached enzymatically and characterized by MALDI-MS as their phenylhydrazone derivatives.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
Phenylhydrazine, 97%