MilliporeSigma
Ruthenium-catalyzed mild C-H oxyfunctionalization of cyclic steroidal ethers.
The Journal of organic chemistry (2007-06-26)
Jong Seok Lee, Hui Cao, Philip L Fuchs
RÉSUMÉ
Ruthenium-catalyzed site-specific C-H oxyfunctionalization of steroidal ethers with periodate or bromate as terminal oxidants in phosphate buffer provided the acid-sensitive C-16 hydroxy compounds in high yields. Phosphate buffer (pH 7.5) significantly inhibits formation of unwanted side products generated under more acidic reaction conditions. A key example is demonstrated at the 100 g scale.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Ruthenium(III) chloride, Ru content 45-55%