MilliporeSigma
ADAMTS13-mediated thrombolysis of t-PA-resistant occlusions in ischemic stroke in mice.
Blood (2016-03-02)
Frederik Denorme, Friederike Langhauser, Linda Desender, Aline Vandenbulcke, Hanspeter Rottensteiner, Barbara Plaimauer, Olivier François, Tommy Andersson, Hans Deckmyn, Friedrich Scheiflinger, Christoph Kleinschnitz, Karen Vanhoorelbeke, Simon F De Meyer
RÉSUMÉ
Rapid vascular recanalization forms the basis for successful treatment of cerebral ischemia. Currently, tissue plasminogen activator (t-PA) is the only approved thrombolytic drug for ischemic stroke. However, t-PA does not always result in efficient thrombus dissolution and subsequent blood vessel recanalization. To better understand thrombus composition, we analyzed thrombi retrieved from ischemic stroke patients and found a distinct presence of von Willebrand factor (VWF) in various samples. Thrombi contained on average 20.3% ± 10.1% VWF, and this was inversely correlated with thrombus red blood cell content. We hypothesized that ADAMTS13 can exert a thrombolytic effect in VWF-containing thrombi in the setting of stroke. To test this, we generated occlusive VWF-rich thrombi in the middle cerebral artery (MCA) of mice. Infusion of t-PA did not dissolve these MCA occlusions. Interestingly, administration of ADAMTS13 5 minutes after occlusion dose-dependently dissolved these t-PA-resistant thrombi resulting in fast restoration of MCA patency and consequently reduced cerebral infarct sizes (P < .005). Delayed ADAMTS13 administration 60 minutes after occlusion was still effective but to a lesser extent (P < .05). These data show for the first time a potent thrombolytic activity of ADAMTS13 in the setting of stroke, which might become useful in treatment of acute ischemic stroke.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Eosin Y Solution, Aqueous