- Highly enantioselective azadiene Diels-Alder reactions catalyzed by chiral N-heterocyclic carbenes.
Highly enantioselective azadiene Diels-Alder reactions catalyzed by chiral N-heterocyclic carbenes.
Journal of the American Chemical Society (2006-06-29)
Ming He, Justin R Struble, Jeffrey W Bode
PMID16802805
RÉSUMÉ
Highly enantioselective, N-heterocyclic carbene (NHC)-catalyzed aza-Diels-Alder reactions are described. A novel chiral triazolium salt based on the cis-1,2-aminoindanol platform serves as an efficient precatalyst for the NHC-catalyzed redox generation of enolate dienophiles that undergo LUMOdiene-controlled Diels-Alder reactions with N-sulfonyl-alpha,beta-unsaturated imines in good yields and with exceptional diastereo- and enantioselectivities (>99% ee). In contrast to uncatalyzed variants, this organocatalytic process proceeds at room temperature without stoichiometric reagents, producing synthetically valuable, enantiomerically pure cis-3,4-disubstituted dihydropyridinone products.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
6,7-Dihydro-2-pentafluorophenyl-5H-pyrrolo[2,1-c]-1,2,4-triazolium tetrafluoroborate, 97%