MilliporeSigma
Natural sweet macromolecules: how sweet proteins work.
Cellular and molecular life sciences : CMLS (2006-07-01)
P A Temussi
RÉSUMÉ
A few proteins, discovered mainly in tropical fruits, have a distinct sweet taste. These proteins have played an important role towards a molecular understanding of the mechanisms of taste. Owing to the huge difference in size, between most sweeteners and sweet proteins, it was believed that they must interact with a different receptor from that of small molecular weight sweeteners. Recent modelling studies have shown that the single sweet taste receptor has multiple active sites and that the mechanism of interaction of sweet proteins is intrinsically different from that of small sweeteners. Small molecular weight sweeteners occupy small receptor cavities inside two subdomains of the receptor, whereas sweet proteins can interact with the sweet receptor according to a mechanism called the 'wedge model' in which they bind to a large external cavity. This review describes these mechanisms and outlines a history of sweet proteins.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Thaumatin from Thaumatococcus daniellii