MilliporeSigma
  • Nerve growth factor and epidermal growth factor rescue PC12 cells from programmed cell death induced by etoposide: distinct modes of protection against cell death by growth factors and a protein-synthesis inhibitor.
Nerve growth factor and epidermal growth factor rescue PC12 cells from programmed cell death induced by etoposide: distinct modes of protection against cell death by growth factors and a protein-synthesis inhibitor.
Neuroscience letters (1994-08-01)
M Nakajima, K Kashiwagi, J Ohta, S Furukawa, K Hayashi, T Kawashima, Y Hayashi
RÉSUMÉ
A rat pheochromocytoma cell line (PC12 cells) died within 24 h in the presence of etoposide (1-40 micrograms/ml), an inhibitor of topoisomerase II. This cytotoxic effect was prevented by either nerve growth (NGF) or epidermal growth factor (EGF). Cycloheximide and actinomycin D also suppressed the cell death as well. Furthermore, a difference among protective modes against etoposide-induced death by growth factors and a protein-synthesis inhibitor was observed: the protective effect of either NGF or EGF remained rather constant as a function of incubation time with etoposide whereas that of cycloheximide declined. These results indicate that etoposide induces programmed death in PC12 cells and that prevention of the programmed cell death by both NGF and EGF is mainly due to inactivation of molecules involved in the death processes rather than suppression of specific protein and/or mRNA synthesis.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Actinomycin D, Ready Made Solution, from Streptomyces sp., 2 mg/mL in DMSO