MilliporeSigma
  • Determination of amino acid enantiopurity and absolute configuration: synergism between configurationally labile metal-based receptors and dynamic covalent interactions.
Determination of amino acid enantiopurity and absolute configuration: synergism between configurationally labile metal-based receptors and dynamic covalent interactions.
Chemistry (Weinheim an der Bergstrasse, Germany) (2013-11-01)
Francesca A Scaramuzzo, Giulia Licini, Cristiano Zonta
RÉSUMÉ
Reliable determination of the enantiomeric excess of free amino acids can be obtained by measuring the induced circular dichroism of a multicomponent assembly formed by a modified tris(2-pyridylmethyl)amine ligand, a zinc salt, and the amino acid of interest. The systems furnish reliable information for all natural amino acids.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
6-Bromo-2-pyridinecarboxaldehyde, 97%