MilliporeSigma
Efficient and selective Al-catalyzed alcohol oxidation via Oppenauer chemistry.
Journal of the American Chemical Society (2006-09-28)
Christopher R Graves, Bi-Shun Zeng, SonBinh T Nguyen
RÉSUMÉ
A highly active and selective Al-based catalytic Oppenauer (O) oxidation is reported. Quantitative and selective oxidations of a variety of benzylic, propargylic, allylic, and aliphatic primary and secondary alcohols were achieved using nitrobenzaldehyde derivatives as the oxidant and simple aluminum compounds as precatalysts.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Trimethylaluminum solution, 2.0 M in toluene