MilliporeSigma
Selective efficacy of depsipeptide in a xenograft model of Epstein-Barr virus-positive lymphoproliferative disorder.
Journal of the National Cancer Institute (2004-10-07)
Sameek Roychowdhury, Robert A Baiocchi, Srinivas Vourganti, Darshna Bhatt, Bradley W Blaser, Aharon G Freud, Jason Chou, Chang-Shi Chen, Jim J Xiao, Mark Parthun, Kenneth K Chan, Charles F Eisenbeis, Amy K Ferketich, Michael R Grever, Ching-Shih Chen, Michael A Caligiuri
RÉSUMÉ
Immune-compromised individuals are at increased risk for developing aggressive Epstein-Barr virus (EBV)-associated lymphoproliferative disorders after primary EBV infection or for reactivation of a preexisting latent EBV infection. We evaluated the effect of depsipeptide, a histone deacetylase inhibitor, on EBV-positive lymphoblastoid cell lines (LCLs) and Burkitt lymphoma cell lines in a mouse model and explored its mechanism of action in vitro. We studied EBV-transformed LCLs, which express a latent III (Lat-III) viral gene profile, as do some EBV-positive lymphoproliferative malignancies, and Burkitt lymphoma cell lines, which express a Lat-I viral gene profile. Cell lines were used to characterize depsipeptide-induced apoptosis, which was evaluated by flow cytometry. Flow cytometry, western blot analyses, and histone deacetylase inhibitors were used to investigate components of prodeath and survival pathways in vitro. We studied depsipeptide's effects on survival with a mouse xenograft model of EBV-positive human B-cell tumors (groups of 10 mice). All statistical tests were two-sided. Depsipeptide (5 mg/m2 of body surface area) treatment was associated with statistically significantly improved survival of mice carrying Lat-III EBV-positive LCL tumors, compared with that of control-treated mice (day 30: for depsipeptide-treated mice, 90% survival, 95% confidence interval [CI] = 73.2% to 100%; for control-treated mice, 20% survival, 95% CI = 5.79% to 69.1%; P<.001), but it was not associated with survival of mice carrying Lat-I EBV-positive Burkitt lymphoma tumors. Depsipeptide induced apoptosis in 64% of LCLs and in 14% of EBV-positive Burkitt lymphoma cells in vitro. Depsipeptide-treated LCL cultures had two distinct cell populations--one sensitive and one resistant to depsipeptide. Depsipeptide-mediated apoptosis was associated with a 12-fold increased level of active caspase 3, but some apoptosis persisted despite z-VAD-fmk treatment to inhibit caspase activity. Depsipeptide-resistant LCLs expressed higher levels of latent membrane protein 1 (LMP1; P = .017), BCL2 (P = .032), and nuclear factor kappaB (NF-kappaB) (P<.001) than depsipeptide-sensitive LCLs; this resistance was circumvented by treatment with PS-1145, an inhibitor of NF-kappaB activation (P<.001). Apoptosis is induced by depsipeptide via caspase-dependent and -independent pathways in Lat-III EBV-positive LCLs and is enhanced by inhibiting NF-kappaB activity. Depsipeptide as a treatment for Lat-III EBV-associated lymphoproliferative disorders should be explored further in clinical trials.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
PS-1145 dihydrochloride, ≥98% (HPLC), solid