MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Chemerin C9 peptide induces receptor internalization through a clathrin-independent pathway.
Acta pharmacologica Sinica (2014-03-25)
Jun-xian Zhou, Dan Liao, Shuo Zhang, Ni Cheng, Hui-qiong He, Richard D Ye
RÉSUMÉ
The chemerin receptor CMKLR1 is one type of G protein-coupled receptors abundant in monocyte-derived dendritic cells and macrophages, which plays a key role in the entry of a subset of immunodeficiency viruses including HIV/SIV into lymphocytes and macrophages. The aim of this work was to investigate how CMKLR1 was internalized and whether its internalization affected cell signaling in vitro. Rat basophilic leukemia RBL-2H3 cells, HEK 293 cells, and HeLa cells were used. CMKLR1 internalization was visualized by confocal microscopy imaging or using a FACScan flow cytometer. Six potential phosphorylation sites (Ser337, Ser343, Thr352, Ser344, Ser347, and Ser350) in CMKLR1 were substituted with alanine using site-directed mutagenesis. Heterologous expression of wild type and mutant CMKLR1 allowed for functional characterization of endocytosis, Ca(2+) flux and extracellular signal-regulated kinase (ERK) phosphorylation. Chemerin and the chemerin-derived nonapeptide (C9) induced dose-dependent loss of cell surface CMKLR1-GFP fusion protein and increased its intracellular accumulation in HEK 293 cells and RBL-2H3 cells stably expressing CMKLR1. Up to 90% of CMKLR1 was internalized after treatment with C9 (1 μmol/L). By using different agents, it was demonstrated that clathrin-independent mechanism was involved in CMKLR1 internalization. Mutations in Ser343 for G protein-coupled receptor kinase phosphorylation and in Ser347 for PKC phosphorylation abrogated CMKLR1 internalization. Loss of CMKLR1 internalization partially enhanced the receptor signaling, as shown by increased Ca(2+) flux and a shorter latency to peak level of ERK phosphorylation. CMKLR1 internalization occurs in a clathrin-independent manner, which negatively regulated the receptor-mediated Ca(2+) flux and ERK phosphorylation.