MilliporeSigma
HSV-1-induced chemokine expression via IFI16-dependent and IFI16-independent pathways in human monocyte-derived macrophages.
Herpesviridae (2012-10-16)
Stine Søby, Rune R Laursen, Lars Østergaard, Jesper Melchjorsen
RÉSUMÉ
Innate recognition is essential in the antiviral response against infection by herpes simplex virus (HSV). Chemokines are important for control of HSV via recruitment of natural killer cells, T lymphocytes, and antigen-presenting cells. We previously found that early HSV-1-mediated chemokine responses are not dependent on TLR2 and TLR9 in human macrophages. Here, we investigated the role of the recently identified innate IFN-inducible DNA receptor IFI16 during HSV-1 infection in human macrophages. Peripheral blood mononuclear cells were purified from buffy coats and monocytes were differentiated to macrophages. Macrophages infected with HSV-1 were analyzed using siRNA-mediated knock-down of IFI16 by real-time PCR, ELISA, and Western blotting. We determined that both CXCL10 and CCL3 are induced independent of HSV-1 replication. IFI16 mediates CCL3 mRNA accumulation during early HSV-1 infection. In contrast, CXCL10 was induced independently of IFI16. Our data provide the first evidence of HSV-1-induced innate immune responses via IFI16 in human primary macrophages. In addition, the data suggest that at least one additional unidentified receptor or innate sensing mechanism is involved in recognizing HSV-1 prior to viral replication.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
MISSION® esiRNA, targeting human IFI16