MilliporeSigma
Nuclear envelope remnants: fluid membranes enriched in sterols and polyphosphoinositides.
PloS one (2009-01-24)
Marie Garnier-Lhomme, Richard D Byrne, Tina M C Hobday, Stephen Gschmeissner, Rudiger Woscholski, Dominic L Poccia, Erick J Dufourc, Banafshé Larijani
RÉSUMÉ
The cytoplasm of eukaryotic cells is a highly dynamic compartment where membranes readily undergo fission and fusion to reorganize the cytoplasmic architecture, and to import, export and transport various cargos within the cell. The double membrane of the nuclear envelope that surrounds the nucleus, segregates the chromosomes from cytoplasm and regulates nucleocytoplasmic transport through pores. Many details of its formation are still unclear. At fertilization the sperm devoid of nuclear envelope pores enters the egg. Although most of the sperm nuclear envelope disassembles, remnants of the envelope at the acrosomal and centriolar fossae do not and are subsequently incorporated into the newly forming male pronuclear envelope. Remnants are conserved from annelid to mammalian sperm. Using lipid mass spectrometry and a new application of deuterium solid-state NMR spectroscopy we have characterized the lipid composition and membrane dynamics of the sperm nuclear envelope remnants in isolated sperm nuclei. We report nuclear envelope remnants are relatively fluid membranes rich in sterols, devoid of sphingomyelin, and highly enriched in polyphosphoinositides and polyunsaturated phospholipids. The localization of the polybasic effector domain of MARCKS illustrates the non-nuclear aspect of the polyphosphoinositides. Based on their atypical biophysical characteristics and phospholipid composition, we suggest a possible role for nuclear envelope remnants in membrane fusion leading to nuclear envelope assembly.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Avanti
12:0 PA, 1,2-dilauroyl-sn-glycero-3-phosphate (sodium salt), powder
Le tarif et la disponibilité ne sont pas disponibles actuellement.