MilliporeSigma
  • Comparison of the ocular tolerability of a latanoprost cationic emulsion versus conventional formulations of prostaglandins: an in vivo toxicity assay.
Comparison of the ocular tolerability of a latanoprost cationic emulsion versus conventional formulations of prostaglandins: an in vivo toxicity assay.
Molecular vision (2009-08-28)
Hong Liang, Christophe Baudouin, Marie-Odile Faure, Grégory Lambert, Françoise Brignole-Baudouin
RÉSUMÉ
Using an established rabbit toxicological model, this in vivo study compared the ocular cytotoxicity of four topical intraocular pressure (IOP)-lowering agents: the commercial benzalkonium chloride (BAC)-containing solutions of 0.005% latanoprost, 0.004% travoprost, 0.03% bimatoprost (containing 0.02%, 0.015%, and 0.005% BAC, respectively), and 0.005% latanoprost in a new cationic emulsion (LCEm) formulation. Thirty adult male New Zealand albino rabbits were used in this study. They were randomly divided into five groups: 50 microl of sterile phosphate-buffered saline (PBS) along with each formulation was applied onto rabbit eyes 15 times at 5 min intervals. The ocular surface changes were investigated using slit-lamp examination, corneal in vivo confocal microscopy (IVCM) for cornea, limbus, conjunctiva/conjunctiva-associated lymphoid tissue (CALT) investigations, and conjunctival imprints for cytology and flow cytometry (FCM) analyses. Antiglaucoma eye drops induced an ocular surface cytotoxicity primarily related to the concentration of their common BAC preservative ((0.02%BAC+)latanoprost> (0.015%BAC+)travoprost> (0.005%BAC+)bimatoprost). LCEm did not induce any obvious signs of toxicity on the rabbit ocular surface with results similar to those of PBS; moreover, the conjunctiva/CALT and cornea had almost normal aspects. These in vivo and ex vivo toxicological procedures performed in an acute stress model confirmed the ocular surface cytotoxicity of BAC-containing antiglaucomatous eye drop solutions. The new formulation, LCEm, was well tolerated without inducing ocular surface damage or CALT activation. The cationic emulsion of latanoprost will most likely have fewer long-term adverse effects on the ocular surface than formulations containing toxic preservative BAC and may improve long-term tolerance over BAC-containing antiglaucomatous topical treatments.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
17-Phenyl-tri-norprostaglandin F-ethyl amide, ≥95%, solid