MilliporeSigma
Cisplatin-induced expression of Gb3 enables verotoxin-1 treatment of cisplatin resistance in malignant pleural mesothelioma cells.
British journal of cancer (2009-12-17)
D Johansson, C Andersson, J Moharer, A Johansson, P Behnam-Motlagh
RÉSUMÉ
A major problem with cisplatin treatment is the development of acquired-drug resistance of the tumour cells. Verotoxin-1 (VT-1) exerts its cytotoxicity by targeting the membrane glycolipid globotriasosylceramide (Gb3), a molecule associated with drug resistance. Cisplatin- and VT-1-induced apoptosis involves mitogen-activated protein kinase (MAPK) activation, and deactivation of MAPKs is associated with cisplatin resistance. This study aimed to investigate whether a sub-toxic concentration of VT-1 could enhance cisplatin-induced apoptosis and overcome acquired-cisplatin resistance in cultured cancer cell lines. P31 and H1299 cells with corresponding cisplatin-resistant sub-lines (P31res/H1299res) were incubated with VT-1 and/or cisplatin followed by determination of Gb3 expression, cell viability, apoptosis, and signalling pathways. Cells from the resistant sub-lines had elevated Gb3 expression compared with the parental cell lines, and cisplatin further increased Gb3 expression, whereas VT-1 reduced the percentage of Gb3-expressing cells. Combination of cisplatin and sub-toxic concentrations of VT-1 led to a super-additive increase of cytotoxicity and TUNEL staining, especially in the cisplatin-resistant sub-lines. Blockade of Gb3 synthesis by a Gb3 synthesis inhibitor not only led to eradicated TUNEL staining of P31 cells, but also sensitised P31res cells to the induction of apoptosis by cisplatin alone. Cisplatin- and VT-1-induced apoptosis involved the MAPK pathways with increased C-Jun N-terminal kinase and MAPK kinase-3 and -6 phosphorylation. We show the presence of Gb3 in acquired-cisplatin resistance in P31res and H1299res cells. Cisplatin up-regulated Gb3 expression in all cells and thus sensitised the cells to VT-1-induced cytotoxicity. A strong super-additive effect of combined cisplatin and a sub-toxic concentration of VT-1 in cisplatin-resistant malignant pleural mesothelioma cells were observed, indicating a new potential clinical-treatment approach.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
DL-threo-1-Phenyl-2-palmitoylamino-3-morpholino-1-propanol, 98%