MilliporeSigma
Synthesis and optoelectronic properties of 2,6-bis(2-anilinoethynyl)pyridine scaffolds.
Chemical science (2012-04-01)
Jeffrey M Engle, Calden N Carroll, Darren W Johnson, Michael M Haley
RÉSUMÉ
A series of sixteen bisphenylureas based on a 2,6-bis(2-anilinoethynyl)pyridine scaffold have been synthesized for use as potential anion sensors. Prior work with one of these receptors revealed a distinct fluorescence response in the presence of a suitable anion source with specificity towards chloride anion. This study demonstrates that the fluorescent properties of these receptors can be tuned through the systematic variation of the pendant functional groups.