MilliporeSigma
N-trifluoromethylthiosaccharin: an easily accessible, shelf-stable, broadly applicable trifluoromethylthiolating reagent.
Angewandte Chemie (International ed. in English) (2014-07-22)
Chunfa Xu, Bingqing Ma, Qilong Shen
RÉSUMÉ
A new, electrophilic trifluoromethylthiolating reagent, N-trifluoromethylthiosaccharin, was developed and can be synthesized in two steps from saccharin within 30 minutes. N-trifluoromethylthiosaccharin is a powerful trifluoromethylthiolating reagent and allows the trifluoromethylthiolation of a variety of nucleophiles such as alcohols, amines, thiols, electron-rich arenes, aldehydes, ketones, acyclic β-ketoesters, and alkynes under mild reaction conditions.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
1-Ethynyl-3-fluorobenzene, 98%