MilliporeSigma
The FEZ1 gene at chromosome 8p22 encodes a leucine-zipper protein, and its expression is altered in multiple human tumors.
Proceedings of the National Academy of Sciences of the United States of America (1999-03-31)
H Ishii, R Baffa, S I Numata, Y Murakumo, S Rattan, H Inoue, M Mori, V Fidanza, H Alder, C M Croce
RÉSUMÉ
Alterations of human chromosome 8p occur frequently in many tumors. We identified a 1.5-Mb common region of allelic loss on 8p22 by allelotype analysis. cDNA selection allowed isolation of several genes, including FEZ1. The predicted Fez1 protein contained a leucine-zipper region with similarity to the DNA-binding domain of the cAMP-responsive activating-transcription factor 5. RNA blot analysis revealed that FEZ1 gene expression was undetectable in more than 60% of epithelial tumors. Mutations were found in primary esophageal cancers and in a prostate cancer cell line. Transcript analysis from several FEZ1-expressing tumors revealed truncated mRNAs, including a frameshift. Alteration and inactivation of the FEZ1 gene may play a role in various human tumors.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Anti-LZTS1 antibody produced in rabbit, IgG fraction of antiserum