MilliporeSigma
  • Gene expression profiles induced by E6 from non-European HPV18 variants reveals a differential activation on cellular processes driving to carcinogenesis.
Gene expression profiles induced by E6 from non-European HPV18 variants reveals a differential activation on cellular processes driving to carcinogenesis.
Virology (2012-06-30)
Verónica Fragoso-Ontiveros, Rosa María Alvarez-García, Adriana Contreras-Paredes, Felipe Vaca-Paniagua, Luis Alonso Herrera, Cesar López-Camarillo, Nadia Jacobo-Herrera, Marcela Lizano-Soberón, Carlos Pérez-Plasencia
RÉSUMÉ
Cervical cancer in developed countries remains as a major concern on public health policies due to incidence and mortality rates. Persistent infection with high risk human papillomavirus is a necessary etiological agent in the progression to invasive cervical carcinoma. A proposed hypothesis is the association between more aggressive HPV variants and the risk to develop cervical cancer. In order to have a global perspective in terms of cellular transcripts and molecular pathways affected by HPV18 E6 intratype variants; we conducted a genome wide analysis of gene expression. Our results show that E6 derived from non-European variants are able to up-regulate cellular transcripts associated to the hallmarks of cancer; such as cell cycle, migration, Wnt pathway and mTor signaling. Moreover, we were able to show that HPV18 E6 from African variant had a major effect on cellular processes such as cell cycle and migration as confirmed by functional studies.