MilliporeSigma
  • Protein domain histochemistry (PDH): binding of the carbohydrate recognition domain (CRD) of recombinant human glycoreceptor CLEC10A (CD301) to formalin-fixed, paraffin-embedded breast cancer tissues.
Protein domain histochemistry (PDH): binding of the carbohydrate recognition domain (CRD) of recombinant human glycoreceptor CLEC10A (CD301) to formalin-fixed, paraffin-embedded breast cancer tissues.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society (2013-01-01)
Peter Nollau, Gerrit Wolters-Eisfeld, Naghmeh Mortezai, Anna-Katharina Kurze, Birgit Klampe, Annegret Debus, Maximilian Bockhorn, Axel Niendorf, Christoph Wagener
RÉSUMÉ
Specialized protein domains bind to posttranslational modifications (PTMs) of proteins, such as phosphorylation or glycosylation. When such PTM-binding protein domains are used as analytical tools, the functional states of cells and tissues can be determined with high precision. Here, we describe the use of recombinant CLEC10A (CD301), a human glycoreceptor of the C-type lectin family, for the detection of ligands in sections from formalin-fixed, paraffin-embedded normal and cancerous mammary tissues. A construct, in which part of the carbohydrate recognition domain (CRD) was deleted, was used as a negative control. In comparison to normal mammary glands, a pronounced staining of tumor tissues was observed. Because the construct with the truncated CRD did not show any tissue staining, the binding of the wild-type glycoreceptor can be attributed to its carbohydrate recognition domain. To distinguish our novel approach from immunohistochemistry, we propose the designation "protein domain histochemistry" (PDH).