MilliporeSigma
Potential role of a navigator gene NAV3 in colorectal cancer.
British journal of cancer (2011-12-17)
E Carlsson, A Ranki, L Sipilä, L Karenko, W M Abdel-Rahman, K Ovaska, L Siggberg, U Aapola, R Ässämäki, V Häyry, K Niiranen, M Helle, S Knuutila, S Hautaniemi, P Peltomäki, K Krohn
RÉSUMÉ
The recently described navigator proteins have a multifaceted role in cytoskeletal dynamics. We report here on the relevance of one of them, navigator 3 (NAV3), in colorectal cancer (CRC). We analysed changes in chromosome 12 and NAV3 copy number in CRC/adenoma samples of 59 patients and in 6 CRC cell lines, using fluorescence in situ hybridisation, loss of heterozygosity, and array-CGH. NAV3 target genes were identified by siRNA depletion, expression arrays, and immunohistochemistry. NAV3 deletion and chromosome 12 polysomy were detected in 30 and 70% of microsatellite stability (MSS) carcinomas, in 23 and 30% of adenomas and in four of six CRC cell lines. NAV3 amplification was found in 25% of MSS samples. NAV3 alterations correlated with lymph node metastasis. In normal colon cells, NAV3 silencing induced upregulation of interleukin 23 receptor (IL23R) and gonadotropin releasing hormone receptor. In MSS and microsatellite instability tumours, IL23R immunoreactivity correlated with Dukes' staging and lymph node metastases, whereas nuclear beta-catenin correlated with lymph node metastases only. NAV3 copy number changes are frequent in CRC and in adenomas, and upregulation of IL23R, following NAV3 silencing, strongly correlates with Dukes' staging and lymph node metastases. This suggests that NAV3 has a role in linking tissue inflammation to cancer development in the colon.