MilliporeSigma
Spinster 2, a sphingosine-1-phosphate transporter, plays a critical role in inflammatory and autoimmune diseases.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2015-09-02)
Michael S Donoviel, Nitai C Hait, Subramaniam Ramachandran, Michael Maceyka, Kazuaki Takabe, Sheldon Milstien, Tamas Oravecz, Sarah Spiegel
RÉSUMÉ
Sphingosine 1-phosphate (S1P) is a pleiotropic bioactive sphingolipid metabolite that regulates numerous processes important for immune responses. S1P is made within cells and must be transported out of cells to exert its effects through activation of 5 specific cell surface GPCRs in an autocrine or paracrine fashion. Spinster 2 (Spns2) transports S1P out of cells, and its deletion in mice reduces circulating levels of S1P, alters immune cell trafficking, and induces lymphopenia. Here we examined the effects of Spns2 deletion on adaptive immune responses and in autoimmune disease models. Airway inflammation and hypersensitivity as well as delayed-type contact hypersensitivity were attenuated in Spns2(-/-) mice. Similarly, Spns2 deletion reduced dextran sodium sulfate- and oxazolone-induced colitis. Intriguingly, Spns2(-/-) mice were protected from the development of experimental autoimmune encephalopathy, a model of the autoimmune disease multiple sclerosis. Deletion of Spns2 also strongly alleviated disease development in collagen-induced arthritis. These results point to a broad role for Spns2-mediated S1P transport in the initiation and development of adaptive immune related disorders.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Fluorure de phénylméthanesulfonyle, ≥98.5% (GC)
Sigma-Aldrich
Fluorure de phénylméthanesulfonyle, ≥99.0% (T)