MilliporeSigma
  • The closed conformation of the LDL receptor is destabilized by the low Ca(++) concentration but favored by the high Mg(++) concentration in the endosome.
The closed conformation of the LDL receptor is destabilized by the low Ca(++) concentration but favored by the high Mg(++) concentration in the endosome.
FEBS letters (2015-11-04)
Juan Martínez-Oliván, Xabier Arias-Moreno, Ramón Hurtado-Guerrero, José Alberto Carrodeguas, Laura Miguel-Romero, Alberto Marina, Pierpaolo Bruscolini, Javier Sancho
RÉSUMÉ
The LDL receptor (LDLR) internalizes LDL and VLDL particles. In the endosomes, it adopts a closed conformation important for recycling, by interaction of two modules of the ligand binding domain (LR4-5) and a β-propeller motif. Here, we investigate by SPR the interactions between those two modules and the β-propeller. Our results indicate that the two modules cooperate to bind the β-propeller. The binding is favored by low pH and by high [Ca(++)]. Our data show that Mg(++), at high concentration in the endosome, favors the formation of the closed conformation by replacing the structuring effect of Ca(++) in LR5. We propose a sequential model of LDL release where formation of the close conformation follows LDL release.