MilliporeSigma
drp, a novel protein expressed at high cell density but not during growth arrest.
DNA and cell biology (1998-06-17)
J E Deyo, P J Chiao, M A Tainsky
RÉSUMÉ
Contact is a vital mechanism used by cells to interact with their environment. Contact with living and nonliving elements adjacent to a cell is the basis for many common biological events ranging from growth regulation to metastasis to embryonic pattern formation. We describe the cloning and characterization of a novel density-regulated protein (drp) whose expression is increased in cultured cells at high density compared with cells at low density. A drp cDNA was isolated from the human teratocarcinoma cell line PA-1. Northern analysis with a drp probe revealed transcripts of 2.8 and 3.2 kb. The drp RNA was expressed in a variety of tissues, with the highest amounts in skeletal and cardiac muscle. Using antipeptide antisera, increasing amounts of a 70-kDa protein were detected using several experimental approaches in several cells lines as cell density is increased. Conditioned medium from high-density cells was unable to induce expression of drp in cells growing at low density. Similarly, growth arrest by serum starvation or transforming growth factor-beta (TGF-beta) treatment failed to elicit drp expression. We conclude that drp is a novel protein whose expression is increased at high cell density but not growth arrest.