MilliporeSigma
Comprehensive molecular pathology analysis of small bowel adenocarcinoma reveals novel targets with potential for clinical utility.
Oncotarget (2015-09-01)
Muhammad A Alvi, Darragh G McArt, Paul Kelly, Marc-Aurel Fuchs, Matthew Alderdice, Clare M McCabe, Victoria Bingham, Claire McGready, Shailesh Tripathi, Frank Emmert-Streib, Maurice B Loughrey, Stephen McQuaid, Perry Maxwell, Peter W Hamilton, Richard Turkington, Jacqueline A James, Richard H Wilson, Manuel Salto-Tellez
RÉSUMÉ
Small bowel accounts for only 0.5% of cancer cases in the US but incidence rates have been rising at 2.4% per year over the past decade. One-third of these are adenocarcinomas but little is known about their molecular pathology and no molecular markers are available for clinical use. Using a retrospective 28 patient matched normal-tumor cohort, next-generation sequencing, gene expression arrays and CpG methylation arrays were used for molecular profiling. Next-generation sequencing identified novel mutations in IDH1, CDH1, KIT, FGFR2, FLT3, NPM1, PTEN, MET, AKT1, RET, NOTCH1 and ERBB4. Array data revealed 17% of CpGs and 5% of RNA transcripts assayed to be differentially methylated and expressed respectively (p < 0.01). Merging gene expression and DNA methylation data revealed CHN2 as consistently hypermethylated and downregulated in this disease (Spearman -0.71, p < 0.001). Mutations in TP53 which were found in more than half of the cohort (15/28) and Kazald1 hypomethylation were both were indicative of poor survival (p = 0.03, HR = 3.2 and p = 0.01, HR = 4.9 respectively). By integrating high-throughput mutational, gene expression and DNA methylation data, this study reveals for the first time the distinct molecular profile of small bowel adenocarcinoma and highlights potential clinically exploitable markers.