MilliporeSigma
AICA-ribosiduria: a novel, neurologically devastating inborn error of purine biosynthesis caused by mutation of ATIC.
American journal of human genetics (2004-04-29)
Sandrine Marie, Benedicte Heron, Pierre Bitoun, Therese Timmerman, Georges Van Den Berghe, Marie-Francoise Vincent
RÉSUMÉ
In a female infant with dysmorphic features, severe neurological defects, and congenital blindness, a positive urinary Bratton-Marshall test led to identification of a massive excretion of 5-amino-4-imidazolecarboxamide (AICA)-riboside, the dephosphorylated counterpart of AICAR (also termed "ZMP"), an intermediate of de novo purine biosynthesis. ZMP and its di- and triphosphate accumulated in the patient's erythrocytes. Incubation of her fibroblasts with AICA-riboside led to accumulation of AICAR, not observed in control cells, suggesting impairment of the final steps of purine biosynthesis, catalyzed by the bifunctional enzyme AICAR transformylase/IMP cyclohydrolase (ATIC). AICAR transformylase was profoundly deficient, whereas the IMP cyclohydrolase level was 40% of normal. Sequencing of ATIC showed a K426R change in the transformylase region in one allele and a frameshift in the other. Recombinant protein carrying mutation K426R completely lacks AICAR transformylase activity.