MilliporeSigma
Leukotriene A4 hydrolase: selective abrogation of leukotriene B4 formation by mutation of aspartic acid 375.
Proceedings of the National Academy of Sciences of the United States of America (2002-03-28)
Peter C Rudberg, Fredrik Tholander, Marjolein M G M Thunnissen, Bengt Samuelsson, Jesper Z Haeggstrom
RÉSUMÉ
Leukotriene A4 (LTA4, 5S-trans-5,6-oxido-7,9-trans-11,14-cis-eicosatetraenoic acid) hydrolase (LTA4H)/aminopeptidase is a bifunctional zinc metalloenzyme that catalyzes the final and rate-limiting step in the biosynthesis of leukotriene B4 (LTB4, 5S,12R-dihydroxy-6,14-cis-8,10-trans-eicosatetraenoic acid), a classical chemoattractant and immune modulating lipid mediator. Two chemical features are key to the bioactivity of LTB4, namely, the chirality of the 12R-hydroxyl group and the cis-trans-trans geometry of the conjugated triene structure. From the crystal structure of LTA4H, a hydrophilic patch composed of Gln-134, Tyr-267, and Asp-375 was identified in a narrow and otherwise hydrophobic pocket, believed to bind LTA4. In addition, Asp-375 belongs to peptide K21, a previously characterized 21-residue active site-peptide to which LTA4 binds during suicide inactivation. In the present report we used site-directed mutagenesis and x-ray crystallography to show that Asp-375, but none of the other candidate residues, is specifically required for the epoxide hydrolase activity of LTA4H. Thus, mutation of Asp-375 leads to a selective loss of the enzyme's ability to generate LTB4 whereas the aminopeptidase activity is preserved. We propose that Asp-375, possibly assisted by Gln-134, acts as a critical determinant for the stereoselective introduction of the 12R-hydroxyl group and thus the biological activity of LTB4.