MilliporeSigma
  • A genome-wide association study of nephrolithiasis in the Japanese population identifies novel susceptible Loci at 5q35.3, 7p14.3, and 13q14.1.
A genome-wide association study of nephrolithiasis in the Japanese population identifies novel susceptible Loci at 5q35.3, 7p14.3, and 13q14.1.
PLoS genetics (2012-03-08)
Yuji Urabe, Chizu Tanikawa, Atsushi Takahashi, Yukinori Okada, Takashi Morizono, Tatsuhiko Tsunoda, Naoyuki Kamatani, Kenjiro Kohri, Kazuaki Chayama, Michiaki Kubo, Yusuke Nakamura, Koichi Matsuda
RÉSUMÉ
Nephrolithiasis is a common nephrologic disorder with complex etiology. To identify the genetic factor(s) for nephrolithiasis, we conducted a three-stage genome-wide association study (GWAS) using a total of 5,892 nephrolithiasis cases and 17,809 controls of Japanese origin. Here we found three novel loci for nephrolithiasis: RGS14-SLC34A1-PFN3-F12 on 5q35.3 (rs11746443; P = 8.51×10⁻¹², odds ratio (OR) = 1.19), INMT-FAM188B-AQP1 on 7p14.3 (rs1000597; P = 2.16×10⁻¹⁴, OR = 1.22), and DGKH on 13q14.1 (rs4142110; P = 4.62×10⁻⁹, OR = 1.14). Subsequent analyses in 21,842 Japanese subjects revealed the association of SNP rs11746443 with the reduction of estimated glomerular filtration rate (eGFR) (P = 6.54×10⁻⁸), suggesting a crucial role for this variation in renal function. Our findings elucidated the significance of genetic variations for the pathogenesis of nephrolithiasis.