MilliporeSigma
Elevated cleaved caspase-3 is associated with shortened overall survival in several cancer types.
International journal of clinical and experimental pathology (2014-09-10)
Qian Hu, Jing Peng, Weiping Liu, Xiaoli He, Ling Cui, Xinlian Chen, Mei Yang, Hongqian Liu, Shanling Liu, He Wang
RÉSUMÉ
Emerging evidence has indicated that apoptotic cells have a compensatory effect on the proliferation of neighboring cells. Recent studies have shown that apoptotic tumor cells stimulate the repopulation of tumors from a small number of surviving cells by cleaved caspase-3 regulation and elevated tumor cleaved (and thus activated) caspase-3 expression levels predict worse treatment outcomes in cancer patients. The prognostic significance of cleaved caspase-3 should be demonstrated in more human cancer types and larger subjects. Here, we examined the cleaved caspase-3 expression in 367 human tumor samples (gastric cancer: 97 cases, ovarian cancer: 65 cases, cervical cancer: 104 cases; colorectal cancer: 101 cases) with immunohistochemistry (IHC) and the relationship between the expression of cleaved caspase-3 and various clinicopathological factors were also detected. We found that, cleaved caspase-3 was significantly associated with pathological risk factors (P < 0.005) for the studied cancers, such as tumor stage, lymph-node metastasis, differentiation and so on. In univariate and multivariate analysis, patients with high expression of cleaved caspase-3 had a significant shorter overall survival time compared with those with low cleaved caspase-3 expression in gastric cancer (P < 0.001), ovarian cancer (P < 0.001), cervical cancer (P = 0.002), colorectal cancer (P < 0.001) individually and in the patients combined (P < 0.001). Cox regression results suggested cleaved caspase-3 as an independent prognosis predictor for the studied four cancer types. Our study showed cleaved caspase-3 was well correlated to progression, aggressive behaviors in the studied cancer, and implicated it as a potential predictive factor for the prognosis of the four cancer types. It also indicated cleaved caspase-3 as a potential therapeutic target for cancer patients.