MilliporeSigma
Cathepsin B in infiltrated lymph nodes is of prognostic significance for patients with nonsmall cell lung carcinoma.
Cancer (2001-01-09)
B Werle, C Kraft, T T Lah, J Kos, U Schanzenbächer, K Kayser, W Ebert, E Spiess
RÉSUMÉ
Tumor cells require specific proteolytic enzymes for invasion and metastasis, including lysosomal peptidases--cathepsins. Cathepsin B is a lysosomal cysteine peptidase, which appears to play a major role in invasion and metastasis of human tumors. In this study, the authors focused on the possible role of cathepsin B in lymphogenic metastasis by investigating the enzyme localization and its activity in lung tumors and corresponding tumor-infiltrated lymph nodes. Cathepsin B activity was determined in lung tumors, lung parenchyma, and tumor cell-infiltrated and noninfiltrated regional lymph nodes of the same patient. The authors investigated 35 cancer patients suffering from nonsmall cell lung carcinoma. Cathepsin B throughout activity was measured by cleavage of the fluorogenic substrate Z-Arg-Arg-AMC at pH 6.0. The median specific cathepsin B activity was highest in tumors, followed by the infiltrated lymph nodes, noninfiltrated lymph nodes, and lung parenchyma. The authors showed a significant 1.8-fold increase in cathepsin B activity in tumor-infiltrated lymph nodes compared with noninfiltrated regional lymph nodes and a 4.5-fold increase in lung tumor tissue compared with lung parenchyma. High cathepsin B activity, both in tumors and tumor cell-infiltrated lymph nodes, indicated poor prognosis for overall survival. Immunohistochemical analysis showed the presence of cathepsin B in histiocytes and tumor cells but not in lymphocytes of lymph node tissue. The authors' findings on higher cathepsin B levels in tumor cell-infiltrated lymph nodes show that increased level of cathepsin B activity is characteristic of the invasive tumor cell phenotype. This corroborates the hypothesis, that tumor cell associated cathepsin B may play a role in lymphogenic metastasis. The authors' results support the use of lymph node associated cathepsin B as a prognostic factor for survival of patients with lung carcinoma.