MilliporeSigma
SIRPA, VCAM1 and CD34 identify discrete lineages during early human cardiovascular development.
Stem cell research (2014-06-27)
Rhys J P Skelton, Magdaline Costa, David J Anderson, Freya Bruveris, Ben W Finnin, Katerina Koutsis, Deevina Arasaratnam, Anthony J White, Arash Rafii, Elizabeth S Ng, Andrew G Elefanty, Edouard G Stanley, Colin W Pouton, John M Haynes, Reza Ardehali, Richard P Davis, Christine L Mummery, David A Elliott
RÉSUMÉ
The study of human cardiogenesis would benefit from a detailed cell lineage fate map akin to that established for the haematopoietic lineages. Here we sought to define cell lineage relationships based on the expression of NKX2-5 and the cell surface markers VCAM1, SIRPA and CD34 during human cardiovascular development. Expression of NKX2-5(GFP) was used to identify cardiac progenitors and cardiomyocytes generated during the differentiation of NKX2-5(GFP/w) human embryonic stem cells (hESCs). Cardiovascular cell lineages sub-fractionated on the basis of SIRPA, VCAM1 and CD34 expression were assayed for differentiation potential and gene expression. The NKX2-5(pos)CD34(pos) population gave rise to endothelial cells that rapidly lost NKX2-5 expression in culture. Conversely, NKX2-5 expression was maintained in myocardial committed cells, which progressed from being NKX2-5(pos)SIRPA(pos) to NKX2-5(pos)SIRPA(pos)VCAM1(pos). Up-regulation of VCAM1 was accompanied by the expression of myofilament markers and reduced clonal capacity, implying a restriction of cell fate potential. Combinatorial expression of NKX2-5, SIRPA, VCAM1 and CD34 can be used to define discrete stages of cardiovascular cell lineage differentiation. These markers identify specific stages of cardiomyocyte and endothelial lineage commitment and, thus provide a scaffold for establishing a fate map of early human cardiogenesis.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
Monoclonal Anti-PTPNS1 antibody produced in mouse, clone 1D10, purified immunoglobulin, buffered aqueous solution