MilliporeSigma
Preeclampsia is associated with a deficiency of lipoxin A4, an endogenous anti-inflammatory mediator.
Fertility and sterility (2014-05-07)
Zhangye Xu, Feng Zhao, Feng Lin, Huiqiu Xiang, Ni Wang, Duyun Ye, Yinping Huang
RÉSUMÉ
To test whether lipoxin A4 (LXA4) deficiency results in preeclampsia. Prospective experimental study. Patient and animal research facilities. Sprague-Dawley rats. We measured LXA4 and its biosynthetic enzymes, blocked the LXA4 signaling pathway, treated experimental rats with preeclampsia with LXA4, and detected inflammatory factors, FPR2/ALX, and 11β-HSD2 to systematically test whether lack of LXA4 results in preeclampsia. We measured serum levels of LXA4 and inflammatory factors using enzyme-linked immunosorbent assay; detected LXA4 biosynthetic enzymes, inflammatory factors, FPR2/ALX, and 11β-HSD2 mRNA expression using reverse transcriptase-polymerase chain reaction (RT-PCR) and real-time RT-PCR; and localized protein expression using immunohistochemistry. FPR2/ALX and LXA4 and its biosynthetic enzymes were found to be decreased in women with preeclampsia. Replenishing LXA4 improved the symptoms of lipopolysaccharide-induced rats with preeclampsia, while blocking LXA4 signaling resulted in preeclampsia. LXA4 significantly reduced interleukin-6 (IL-6), tumor necrosis factor-α, and IFN-γ but increased IL-10, LXA4 up-regulated 11β-HSD2. A deficiency of LXA4 may result in preeclampsia, which might be ascribed to a reduction in inflammation response, oxidative stress, and regulation of 11β-HSD2.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Monoclonal Anti-FPRL1 antibody produced in mouse, clone 2D8, purified immunoglobulin, buffered aqueous solution