MilliporeSigma
A new mutation in the gene encoding mitochondrial seryl-tRNA synthetase as a cause of HUPRA syndrome.
BMC nephrology (2013-09-17)
Henry Rivera, Elena Martín-Hernández, Aitor Delmiro, María Teresa García-Silva, Pilar Quijada-Fraile, Rafael Muley, Joaquín Arenas, Miguel A Martín, Francisco Martínez-Azorín
RÉSUMÉ
HUPRA syndrome is a rare mitochondrial disease characterized by hyperuricemia, pulmonary hypertension, renal failure in infancy and alkalosis. This syndrome was previously described in three patients with a homozygous mutation c.1169A > G (p.D390G) in SARS2, encoding the mitochondrial seryl-tRNA synthetase. Here we report the clinical and genetic findings in a girl and her brother. Both patients were clinically diagnosed with the HUPRA syndrome. Analysis of the pedigree identified a new homozygous mutation c.1205G > A (p.R402H) in SARS2 gene. This mutation is very rare in the population and it is located at the C-terminal globular domain of the homodimeric enzyme very close to p.D390G. Several data support that p.R402H mutation in SARS2 is a new cause of HUPRA syndrome.