MilliporeSigma
  • Expression of ECRG4, a novel esophageal cancer-related gene, downregulated by CpG island hypermethylation in human esophageal squamous cell carcinoma.
Expression of ECRG4, a novel esophageal cancer-related gene, downregulated by CpG island hypermethylation in human esophageal squamous cell carcinoma.
World journal of gastroenterology (2003-06-12)
Chun-Mei Yue, Da-Jun Deng, Mei-Xia Bi, Li-Ping Guo, Shih-Hsin Lu
RÉSUMÉ
To study the mechanisms responsible for inactivation of a novel esophageal cancer related gene 4 (ECRG4) in esophageal squamous cell carcinoma (ESCC). A pair of primers was designed to amplify a 220 bp fragment, which contains 16 CpG sites in the core promoter region of the ECRG 4 gene. PCR products of bisulfite-modified CpG islands were analyzed by denaturing high-performance liquid chromatography (DHPLC), which were confirmed by DNA sequencing. The methylation status of ECRG 4 promoter in 20 cases of esophageal cancer and the adjacent normal tissues, 5 human tumor cell lines (esophageal cancer cell line-NEC, EC109, EC9706; gastric cancer cell line- GLC; human embryo kidney cell line-Hek293) and 2 normal esophagus tissues were detected. The expression level of the ECRG 4 gene in these samples was examined by RT-PCR. The expression level of ECRG 4 gene was varied. Of 20 esophageal cancer tissues, nine were unexpressed, six were lowly expressed and five were highly expressed compared with the adjacent tissues and the 2 normal esophageal epithelia. In addition, 4 out of the 5 human cell lines were also unexpressed. A high frequency of methylation was revealed in 12 (8 unexpressed and 4 lowly expressed) of the 15 (80 %) downregulated cancer tissues and 3 of the 4 unexpressed cell lines. No methylation peak was observed in the two highly expressed normal esophageal epithelia and the methylation frequency was low (3/20) among the 20 cases in the highly expressed adjacent tissues. The methylation status of the samples was consistent with the result of DNA sequencing. These results indicate that the inactivation of ECRG 4 gene by hypermethylation is a frequent molecular event in ESCC and may be involved in the carcinogenesis of this cancer.