MilliporeSigma
MST1 mutations in autosomal recessive primary immunodeficiency characterized by defective naive T-cell survival.
Blood (2011-12-17)
Nadine T Nehme, Jana Pachlopnik Schmid, Franck Debeurme, Isabelle André-Schmutz, Annick Lim, Patrick Nitschke, Frédéric Rieux-Laucat, Patrick Lutz, Capucine Picard, Nizar Mahlaoui, Alain Fischer, Geneviève de Saint Basile
RÉSUMÉ
The molecular mechanisms that underlie T-cell quiescence are poorly understood. In the present study, we report a primary immunodeficiency phenotype associated with MST1 deficiency and primarily characterized by a progressive loss of naive T cells. The in vivo consequences include recurrent bacterial and viral infections and autoimmune manifestations. MST1-deficient T cells poorly expressed the transcription factor FOXO1, the IL-7 receptor, and BCL2. Conversely, FAS expression and the FAS-mediating apoptotic pathway were up-regulated. These abnormalities suggest that increased cell death of naive and proliferating T cells is the main mechanism underlying this novel immunodeficiency. Our results characterize a new mechanism in primary T-cell immunodeficiencies and highlight a role of the MST1/FOXO1 pathway in controlling the death of human naive T cells.