MilliporeSigma
Frequencies of HLA-DQ2 and HLA-DQ8 haplotypes in Czech and Slovak coeliac patients and the healthy population.
Acta biochimica Polonica (2014-03-25)
Katerina Wroblova, Michal Kolorz, Igor Pav, Zuzana Horakova, Petra Filipova, Milan Bartos, Ladislava Bartosova
RÉSUMÉ
Coeliac disease is an autoimmune disorder with genetic predisposition. The aim was to determine the frequency of HLA-DQ2 and HLA-DQ8 in Czech and Slovak patients and the healthy population. The study included 127 patients and 66 healthy volunteers. HLA-DQ2 was identified in 85.03% patients, and 24.24% healthy individuals (P=0.0001; OR17.7632; CI=8.4347-37.4088). HLA-DQ8 was identified in 11.81% patients and 15.5% healthy individuals. HLA-DQ8 occurred more often in HLA-DQ2-negative patients compared to HLA-DQ2-positive patients (P=0.0494; OR3.5; CI 1.0428-11.7468). At least one of the studied HLA-variants was found more often in patients than in healthy individuals (P=0.0001; OR58.8; CI 7.6856-449.8602).