MilliporeSigma
PNMA1 promotes cell growth in human pancreatic ductal adenocarcinoma.
International journal of clinical and experimental pathology (2014-08-15)
Shu-Heng Jiang, Ping He, Ming-Ze Ma, Yang Wang, Rong-Kun Li, Fang Fang, Ying Fu, Guang-Ang Tian, Wen-Xin Qin, Zhi-Gang Zhang
RÉSUMÉ
Paraneoplastic Ma1 (PNMA1) is a member of an expanding family of 'brain/testis' proteins involved in an autoimmune disorder defined as paraneoplastic neurological syndrome (PNS). Although it is widely studied in PNS, little is known about the underlying clinical significance and biological function of PNMA1 in tumors. Here, we find that elevated PNMA1 expression is more commonly observed in pancreatic ductal adenocarcinoma (PDAC) cell lines, compared with normal pancreatic cell and tissues from pancreatic ductal adenocarcinoma patient. Besides, higher PNMA1 expression is closely correlated with large tumor size. Suppression of endogenous PNMA1 expression decreases cell viability and promotes cell apoptosis. Subsequent studies reveal that the PI3K/AKT, MAPK/ERK pathway and members of the anti-apoptotic Bcl-2 family may be involved in the pro-survival and anti-apoptotic effect of PNMA1 on PDAC. Taken together, this study provides evidence that PNMA1 is involved in tumor growth of pancreatic carcinoma and PNMA1-related pathways might represent a new treatment strategy.